Assay Details
| Gene Symbol: tmed7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313708.1 | NP_001300637.1 | 2 | 309 | 74 | |
| NM_001313709.1 | NP_001300638.1 | 2 | 306 | 74 | ||
| NM_001313710.1 | NP_001300639.1 | 2 | 218 | 74 | ||
| NM_212912.2 | NP_998077.2 | 2 | 221 | 74 | ||
| GenBank mRNA | BC067383.1 | - | 1 | 147 | 74 | |
| BC159190.1 | - | 2 | 299 | 74 | ||
| CN505306.1 | - | 2 | 326 | 74 | ||
| DT881625.1 | - | 2 | 184 | 74 | ||
| EH456276.1 | - | 2 | 198 | 74 | ||
More Information
Set Membership: |
|

tmed7
Genomic Map