Assay Details
| Gene Symbol: foxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212913.2 | NP_998078.2 | 1 | 1452 | 126 | |
| GenBank mRNA | BC067138.1 | - | 1 | 1453 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

foxd1| Gene Symbol: foxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212913.2 | NP_998078.2 | 1 | 1452 | 126 | |
| GenBank mRNA | BC067138.1 | - | 1 | 1453 | 126 | |
Set Membership: |
|