Assay Details
| Gene Symbol: sox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213118.1 | NP_998283.1 | 1 | 1646 | 165 | |
| GenBank mRNA | AB242329.1 | - | 1 | 1639 | 174 | |
| BC056743.1 | - | 1 | 1648 | 165 | ||
| BC065656.1 | - | 1 | 1646 | 165 | ||
More Information
Set Membership: |
|

sox2
Genomic Map