Assay Details
| Gene Symbol: sox4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213122.2 | NP_998287.1 | 1 | 3151 | 119 | |
| GenBank mRNA | AJ579375.1 | - | 1 | 3145 | 119 | |
| BC056564.1 | - | 1 | 3151 | 119 | ||
| BC065354.1 | - | 1 | 3151 | 119 | ||
More Information
Set Membership: |
|

sox4a
Genomic Map