Assay Details
| Gene Symbol: socs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213304.1 | NP_998469.1 | 2 | 1850 | 111 | |
| GenBank mRNA | BC066573.1 | - | 2 | 1850 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

socs3b| Gene Symbol: socs3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213304.1 | NP_998469.1 | 2 | 1850 | 111 | |
| GenBank mRNA | BC066573.1 | - | 2 | 1850 | 111 | |
Set Membership: |
|