Assay Details
| Gene Symbol: fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214705.1 | NP_999870.1 | 1 | 2252 | 118 | |
| GenBank mRNA | AY391427.1 | - | 1 | 2245 | 118 | |
| BC067169.1 | - | 1 | 2252 | 118 | ||
More Information
Set Membership: |
|

fam43a
Genomic Map