Assay Details
| Gene Symbol: asb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214725.1 | NP_999890.1 | 8 | 2046 | 118 | |
| GenBank mRNA | BC067369.1 | - | 8 | 2046 | 118 | |
| BC167479.1 | - | 7 | 2324 | 113 | ||
More Information
Set Membership: |
|

asb2b
Genomic Map