Assay Details
| Gene Symbol: casp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ812120.1 | - | 4 | 356 | 130 | |
| MG957995.1 | - | 4 | 356 | 130 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

casp3b| Gene Symbol: casp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ812120.1 | - | 4 | 356 | 130 | |
| MG957995.1 | - | 4 | 356 | 130 | ||
Set Membership: |
|