Assay Details
| Gene Symbol: cmtm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002492.1 | NP_001002492.1 | 1 - 2 | 518 | 100 | |
| GenBank mRNA | BC076315.1 | - | 1 - 2 | 518 | 100 | |
| BC152629.1 | - | 1 - 2 | 661 | 100 | ||
More Information
Set Membership: |
|

cmtm4
Genomic Map