Assay Details
| Gene Symbol: nrbf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002613.1 | NP_001002613.1 | 3 - 4 | 259 | 73 | |
| GenBank mRNA | BC075978.1 | - | 3 - 4 | 259 | 73 | |
More Information
Set Membership: |
|

nrbf2b
Genomic Map