Assay Details
| Gene Symbol: bpgm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002630.1 | NP_001002630.1 | 2 - 3 | 658 | 72 | |
| GenBank mRNA | BC075949.1 | - | 2 - 3 | 658 | 72 | |
More Information
Set Membership: |
|

bpgm
Genomic Map