Assay Details
| Gene Symbol: paqr5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003573.2 | NP_001003573.1 | 7 - 8 | 871 | 66 | |
| XM_009303205.2 | XP_009301480.1 | 6 - 7 | 900 | 66 | ||
| GenBank mRNA | BC078202.1 | - | 7 - 8 | 875 | 66 | |
| BC152167.1 | - | 6 - 7 | 821 | 66 | ||
More Information
Set Membership: |
|

paqr5b
Genomic Map