Assay Details
| Gene Symbol: morc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003579.1 | NP_001003579.1 | 3 - 4 | 387 | 79 | |
| GenBank mRNA | BC078200.1 | - | 3 - 4 | 387 | 79 | |
More Information
Set Membership: |
|

morc3b
Genomic Map