Assay Details
| Gene Symbol: tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003781.1 | NP_001003781.1 | 2 - 3 | 181 | 66 | |
| XM_005171838.4 | XP_005171895.1 | 2 - 3 | 315 | 66 | ||
| XM_005171840.4 | XP_005171897.1 | 2 - 3 | 292 | 66 | ||
| GenBank mRNA | BC079504.1 | - | 2 - 3 | 181 | 66 | |
More Information
Set Membership: |
|

tango2
Genomic Map