Assay Details
| Gene Symbol: psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003832.1 | NP_001003832.1 | 4 - 5 | 284 | 75 | |
| GenBank mRNA | AY648827.1 | - | 4 - 5 | 284 | 75 | |
| BC083283.1 | - | 4 - 5 | 294 | 75 | ||
| BC155769.1 | - | 4 - 5 | 291 | 75 | ||
| BC165828.1 | - | 4 - 5 | 295 | 75 | ||
More Information
Set Membership: |
|

psmc6
Genomic Map