Assay Details
| Gene Symbol: cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003838.1 | NP_001003838.1 | 3 - 4 | 385 | 68 | |
| GenBank mRNA | AF393213.1 | - | 3 - 4 | 385 | 68 | |
| BC171621.1 | - | 3 - 4 | 354 | 68 | ||
More Information
Set Membership: |
|

cby1
Genomic Map