Assay Details
| Gene Symbol: tmem144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003999.2 | NP_001003999.1 | 10 - 11 | 1084 | 81 | |
| GenBank mRNA | BC080225.1 | - | 10 - 11 | 1089 | 81 | |
| BC152621.1 | - | 10 - 11 | 1118 | 81 | ||
More Information
Set Membership: |
|

tmem144a
Genomic Map