Assay Details
| Gene Symbol: tmem144a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003999.2 | NP_001003999.1 | 11 - 12 | 1145 | 99 | |
| GenBank mRNA | BC080225.1 | - | 11 - 12 | 1150 | 99 | |
| BC152621.1 | - | 11 - 12 | 1179 | 99 | ||
More Information
Set Membership: |
|

tmem144a
Genomic Map