Assay Details
| Gene Symbol: serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004539.1 | NP_001004539.1 | 4 - 5 | 454 | 72 | |
| GenBank mRNA | BC081582.1 | - | 4 - 5 | 454 | 72 | |
| BC155160.1 | - | 4 - 5 | 460 | 72 | ||
More Information
Set Membership: |
|

serpinf1
Genomic Map