Assay Details
| Gene Symbol: gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004563.1 | NP_001004563.1 | 8 - 9 | 1180 | 64 | |
| GenBank mRNA | BC081629.1 | - | 8 - 9 | 1180 | 64 | |
More Information
Set Membership: |
|

gdap2
Genomic Map