Assay Details
| Gene Symbol: ildr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004642.2 | NP_001004642.2 | 2 - 3 | 455 | 87 | |
| GenBank mRNA | BC078432.1 | - | 2 - 3 | 456 | 87 | |
| CU638769.1 | - | 456 | 87 | |||
| EH440533.1 | - | 2 - 3 | 454 | 87 | ||
More Information
Set Membership: |
|

ildr1b
Genomic Map