Assay Details
| Gene Symbol: tgm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004647.2 | NP_001004647.1 | 5 - 6 | 680 | 78 | |
| GenBank mRNA | BC078420.1 | - | 5 - 6 | 684 | 78 | |
More Information
Set Membership: |
|

tgm2a
Genomic Map