Assay Details
| Gene Symbol: hmgb2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004674.1 | NP_001004674.1 | 3 - 4 | 363 | 56 | |
| GenBank mRNA | BC081415.1 | - | 3 - 4 | 363 | 56 | |
| BC165150.1 | - | 2 - 3 | 335 | 56 | ||
More Information
Set Membership: |
|

hmgb2b
Genomic Map