Assay Details
| Gene Symbol: hpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005769.1 | NP_001005769.1 | 3 - 4 | 439 | 66 | |
| GenBank mRNA | BC055260.1 | - | 3 - 4 | 66 | ||
| BC083292.1 | - | 3 - 4 | 439 | 66 | ||
| BC155158.1 | - | 3 - 4 | 66 | |||
More Information
Set Membership: |
|

hpf1
Genomic Map