Assay Details
| Gene Symbol: epha4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005919.2 | NP_001005919.2 | 16 - 17 | 2930 | 111 | |
| NM_001365171.1 | NP_001352100.1 | 16 - 17 | 2957 | 111 | ||
| XM_005171315.4 | XP_005171372.1 | 17 - 18 | 3349 | 111 | ||
| XM_005171316.4 | XP_005171373.1 | 17 - 18 | 3322 | 111 | ||
| XM_005171317.4 | XP_005171374.1 | 16 - 17 | 3239 | 111 | ||
| XM_005171318.4 | XP_005171375.1 | 16 - 17 | 3213 | 111 | ||
| GenBank mRNA | BC098529.1 | - | 16 - 17 | 2900 | 111 | |
More Information
Set Membership: |
|

epha4a
Genomic Map