Assay Details
| Gene Symbol: epha4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005919.2 | NP_001005919.2 | 7 - 8 | 1682 | 60 | |
| NM_001365171.1 | NP_001352100.1 | 7 - 8 | 1682 | 60 | ||
| XM_005171315.4 | XP_005171372.1 | 7 - 8 | 1966 | 60 | ||
| XM_005171316.4 | XP_005171373.1 | 7 - 8 | 1966 | 60 | ||
| XM_005171317.4 | XP_005171374.1 | 7 - 8 | 1964 | 60 | ||
| XM_005171318.4 | XP_005171375.1 | 7 - 8 | 1965 | 60 | ||
| XM_009298712.3 | XP_009296987.1 | 7 - 8 | 1969 | 60 | ||
| GenBank mRNA | BC098529.1 | - | 7 - 8 | 1677 | 60 | |
| KM655611.1 | - | 7 - 8 | 1600 | 60 | ||
More Information
Set Membership: |
|

epha4a
Genomic Map