Assay Details
| Gene Symbol: glrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005942.1 | NP_001005942.1 | 1 - 2 | 273 | 84 | |
| GenBank mRNA | BC083468.1 | - | 1 - 2 | 273 | 84 | |
| BC164232.1 | - | 1 - 2 | 242 | 84 | ||
More Information
Set Membership: |
|

glrx
Genomic Map