Assay Details
| Gene Symbol: ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005964.1 | NP_001005964.1 | 298 | 79 | ||
| GenBank mRNA | BC083303.1 | - | 298 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ap3s1| Gene Symbol: ap3s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005964.1 | NP_001005964.1 | 298 | 79 | ||
| GenBank mRNA | BC083303.1 | - | 298 | 79 | ||
Set Membership: |
|