Assay Details
| Gene Symbol: atp6v0e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006038.2 | NP_001006038.2 | 2 - 3 | 256 | 70 | |
| GenBank mRNA | BC083377.1 | - | 2 - 3 | 238 | 70 | |
| BC164229.1 | - | 185 | 70 | |||
More Information
Set Membership: |
|

atp6v0e1
Genomic Map