Assay Details
| Gene Symbol: acin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005171275.1 | XP_005171332.1 | 17 - 18 | 5542 | 82 | |
| XM_005171276.1 | XP_005171333.1 | 17 - 18 | 5539 | 82 | ||
| GenBank mRNA | BC116536.1 | - | 11 - 12 | 1415 | 82 | |
| CN504472.1 | - | 2 - 3 | 431 | 82 | ||
| DT078076.1 | - | 5 - 6 | 695 | 82 | ||
| EB961040.1 | - | 1 - 2 | 111 | 82 | ||
| EE691442.1 | - | 2 - 3 | 239 | 82 | ||
More Information
Set Membership: |
|

acin1a
Genomic Map