Assay Details
| Gene Symbol: spega | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007109.1 | NP_001007110.1 | 5 - 6 | 2230 | 97 | |
| GenBank mRNA | AY578914.1 | - | 5 - 6 | 2230 | 97 | |
More Information
Set Membership: |
|

spega
Genomic Map