Assay Details
| Gene Symbol: dnajc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007162.2 | NP_001007163.1 | 6 - 7 | 750 | 63 | |
| XM_005158728.4 | XP_005158785.1 | 6 - 7 | 995 | 63 | ||
| GenBank mRNA | BC162857.1 | - | 6 - 7 | 750 | 63 | |
| BC162873.1 | - | 6 - 7 | 750 | 63 | ||
More Information
Set Membership: |
|

dnajc27
Genomic Map