Assay Details
| Gene Symbol: ptpn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007200.1 | NP_001007201.1 | 19 - 20 | 1987 | 67 | |
| GenBank mRNA | AY590885.1 | - | 1 - 2 | 183 | 67 | |
| BC162720.1 | - | 19 - 20 | 2001 | 67 | ||
More Information
Set Membership: |
|

ptpn4a
Genomic Map