Assay Details
| Gene Symbol: ptpn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007200.1 | NP_001007201.1 | 21 - 22 | 2144 | 84 | |
| GenBank mRNA | AY590885.1 | - | 3 - 4 | 340 | 84 | |
| BC162720.1 | - | 21 - 22 | 2158 | 84 | ||
| FN428714.1 | - | 1 - 2 | 64 | 84 | ||
More Information
Set Membership: |
|

ptpn4a
Genomic Map