Assay Details
| Gene Symbol: ptpn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007200.1 | NP_001007201.1 | 7 - 8 | 602 | 120 | |
| GenBank mRNA | BC162720.1 | - | 7 - 8 | 607 | 120 | |
More Information
Set Membership: |
|

ptpn4a
Genomic Map