Assay Details
| Gene Symbol: anxa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007302.2 | NP_001007303.1 | 5 - 6 | 366 | 73 | |
| GenBank mRNA | BC085679.1 | - | 5 - 6 | 366 | 73 | |
| BC153566.1 | - | 5 - 6 | 319 | 73 | ||
| BC159189.1 | - | 5 - 6 | 304 | 73 | ||
| BC164379.1 | - | 3 - 4 | 282 | 73 | ||
| DV588921.1 | - | 5 - 6 | 365 | 73 | ||
More Information
Set Membership: |
|

anxa3b
Genomic Map