Assay Details
| Gene Symbol: anxa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007302.2 | NP_001007303.1 | 7 - 8 | 568 | 93 | |
| GenBank mRNA | BC085679.1 | - | 7 - 8 | 568 | 93 | |
| BC153566.1 | - | 7 - 8 | 521 | 93 | ||
| BC164379.1 | - | 5 - 6 | 484 | 93 | ||
| DV588921.1 | - | 7 - 8 | 567 | 93 | ||
More Information
Set Membership: |
|

anxa3b
Genomic Map