Assay Details
| Gene Symbol: fosb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328202.1 | NP_001315131.1 | 2 - 3 | 451 | 108 | |
| XM_005173641.4 | XP_005173698.1 | 2 - 3 | 1360 | 108 | ||
| XM_005173642.4 | XP_005173699.1 | 2 - 3 | 1359 | 108 | ||
| GenBank mRNA | BC085664.1 | - | 2 - 3 | 424 | 108 | |
| EH434631.1 | - | 2 - 3 | 460 | 109 | ||
More Information
Set Membership: |
|

fosb
Genomic Map