Assay Details
| Gene Symbol: samm50l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007343.1 | NP_001007344.1 | 3 - 4 | 352 | 63 | |
| GenBank mRNA | BC085536.1 | - | 3 - 4 | 352 | 63 | |
More Information
Set Membership: |
|

samm50l
Genomic Map