Assay Details
| Gene Symbol: eif4ebp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007354.2 | NP_001007355.1 | 2 - 3 | 339 | 102 | |
| GenBank mRNA | BC085595.1 | - | 2 - 3 | 340 | 102 | |
| BC154178.1 | - | 2 - 3 | 375 | 102 | ||
More Information
Set Membership: |
|

eif4ebp3
Genomic Map