Assay Details
| Gene Symbol: nexn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083855.2 | NP_001077324.2 | 2 - 3 | 479 | 122 | |
| GenBank mRNA | BC085519.1 | - | 2 - 3 | 493 | 122 | |
| BC154179.1 | - | 476 | 122 | |||
| DQ464901.1 | - | 1 - 2 | 162 | 122 | ||
More Information
Set Membership: |
|

nexn
Genomic Map