Assay Details
| Gene Symbol: galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007432.2 | NP_001007433.2 | 3 - 4 | 357 | 65 | |
| GenBank mRNA | BC085413.1 | - | 3 - 4 | 357 | 65 | |
| BC165820.1 | - | 3 - 4 | 294 | 65 | ||
More Information
Set Membership: |
|

galk2
Genomic Map