Assay Details
| Gene Symbol: mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007764.1 | NP_001007765.1 | 3 - 4 | 383 | 65 | |
| XM_005168001.4 | XP_005168058.1 | 3 - 4 | 401 | 65 | ||
| GenBank mRNA | BC086509.1 | - | 3 - 4 | 383 | 65 | |
More Information
Set Membership: |
|

mrpl16
Genomic Map