Assay Details
| Gene Symbol: med28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007770.1 | NP_001007771.1 | 2 - 3 | 245 | 79 | |
| GenBank mRNA | BC085569.1 | - | 2 - 3 | 245 | 79 | |
| BC164956.1 | - | 2 - 3 | 266 | 79 | ||
More Information
Set Membership: |
|

med28
Genomic Map