Assay Details
| Gene Symbol: sat2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008598.1 | NP_001008598.1 | 2 - 3 | 188 | 140 | |
| GenBank mRNA | BC086833.1 | - | 2 - 3 | 188 | 140 | |
| BC164882.1 | - | 2 - 3 | 150 | 140 | ||
More Information
Set Membership: |
|

sat2a
Genomic Map