Assay Details
| Gene Symbol: renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126443.1 | NP_001119915.1 | 5 - 6 | 576 | 66 | |
| GenBank mRNA | BC086817.1 | - | 5 - 6 | 617 | 66 | |
More Information
Set Membership: |
|

renbp
Genomic Map