Assay Details
| Gene Symbol: stk35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008627.1 | NP_001008627.1 | 1 - 2 | 628 | 93 | |
| GenBank mRNA | BC086732.1 | - | 1 - 2 | 628 | 93 | |
| BC152611.1 | - | 1 - 2 | 626 | 93 | ||
| BC165829.1 | - | 1 - 2 | 505 | 93 | ||
More Information
Set Membership: |
|

stk35l
Genomic Map