Assay Details
| Gene Symbol: rida | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012315.2 | NP_001012315.1 | 1 - 2 | 190 | 53 | |
| GenBank mRNA | BC079492.1 | - | 1 - 2 | 143 | 53 | |
More Information
Set Membership: |
|

rida
Genomic Map