Assay Details
| Gene Symbol: rybpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012373.2 | NP_001012373.1 | 2 - 3 | 227 | 76 | |
| GenBank mRNA | AY885249.1 | - | 2 - 3 | 155 | 76 | |
| BC125899.1 | - | 2 - 3 | 227 | 76 | ||
| BC165507.1 | - | 2 - 3 | 189 | 76 | ||
More Information
Set Membership: |
|

rybpb
Genomic Map