Assay Details
| Gene Symbol: rybpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012373.2 | NP_001012373.1 | 4 - 5 | 601 | 61 | |
| GenBank mRNA | AY885249.1 | - | 4 - 5 | 529 | 61 | |
| BC125899.1 | - | 4 - 5 | 601 | 61 | ||
| BC165507.1 | - | 4 - 5 | 563 | 61 | ||
More Information
Set Membership: |
|

rybpb
Genomic Map